Learn More
2-Hydroxyphenylacetic acid, 98%
CAS: 614-75-5 | C8H8O3 | 152.149 g/mol
$88.44 - $273.62
Chemical Identifiers
| CAS | 614-75-5 |
|---|---|
| Molecular Formula | C8H8O3 |
| Molecular Weight (g/mol) | 152.149 |
| MDL Number | MFCD00004323 |
| InChI Key | CCVYRRGZDBSHFU-UHFFFAOYSA-N |
| Synonym | 2-hydroxyphenylacetic acid, 2-2-hydroxyphenyl acetic acid, 2-hydroxyphenyl acetic acid, benzeneacetic acid, 2-hydroxy, 2-hydroxybenzeneacetic acid, o-hydroxyphenylacetic acid, acetic acid, o-hydroxyphenyl, 2-hydroxyphenylacetate, o-hydroxyphenyl acetic acid, ortho-hydroxyphenylacetic acid |
| PubChem CID | 11970 |
| ChEBI | CHEBI:28478 |
| IUPAC Name | 2-(2-hydroxyphenyl)acetic acid |
| SMILES | C1=CC=C(C(=C1)CC(=O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1392409
|
Thermo Scientific Chemicals
A1392409 |
10 g |
Each for $88.44
|
|
|||||
|
AAA1392418
|
Thermo Scientific Chemicals
A1392418 |
50 g |
Each for $273.62
|
|
|||||
Description
2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.
Solubility
Soluble in Methanol and Water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 614-75-5 | |
| 152.149 | |
| CCVYRRGZDBSHFU-UHFFFAOYSA-N | |
| 11970 | |
| 2-(2-hydroxyphenyl)acetic acid |
| C8H8O3 | |
| MFCD00004323 | |
| 2-hydroxyphenylacetic acid, 2-2-hydroxyphenyl acetic acid, 2-hydroxyphenyl acetic acid, benzeneacetic acid, 2-hydroxy, 2-hydroxybenzeneacetic acid, o-hydroxyphenylacetic acid, acetic acid, o-hydroxyphenyl, 2-hydroxyphenylacetate, o-hydroxyphenyl acetic acid, ortho-hydroxyphenylacetic acid | |
| CHEBI:28478 | |
| C1=CC=C(C(=C1)CC(=O)O)O |
Specifications
| 614-75-5 | |
| C8H8O3 | |
| 10 g | |
| 2-hydroxyphenylacetic acid, 2-2-hydroxyphenyl acetic acid, 2-hydroxyphenyl acetic acid, benzeneacetic acid, 2-hydroxy, 2-hydroxybenzeneacetic acid, o-hydroxyphenylacetic acid, acetic acid, o-hydroxyphenyl, 2-hydroxyphenylacetate, o-hydroxyphenyl acetic acid, ortho-hydroxyphenylacetic acid | |
| CCVYRRGZDBSHFU-UHFFFAOYSA-N | |
| 2-(2-hydroxyphenyl)acetic acid | |
| 11970 | |
| 152.15 | |
| 2-Hydroxyphenylacetic acid |
| 145°C to 147°C | |
| MFCD00004323 | |
| 908000 | |
| Soluble in Methanol and Water. | |
| C1=CC=C(C(=C1)CC(=O)O)O | |
| 152.149 | |
| CHEBI:28478 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-393-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only