missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyrimidine hydrochloride, 98%

CAS: 38353-09-2 | C4H4N2O·ClH | 132.55 g/mol

$40.16 - $389.33

Chemical Identifiers

CAS 38353-09-2
Molecular Formula C4H4N2O·ClH
Molecular Weight (g/mol) 132.55
MDL Number MFCD00012781
InChI Key IAJINJSFYTZPEJ-UHFFFAOYSA-N
Synonym 2-hydroxypyrimidine hydrochloride, pyrimidin-2-ol hydrochloride, 2 1h-pyrimidinone, monohydrochloride, 2-hydroxypyrimidine hcl, 2-pyrimidinone hydrochloride, pyrimidin-2 1h-one hydrochloride, 2-hydroxypyrimidinehydrochloride, pyrimidin-2-one monohydrochloride, 1,2-dihydropyrimidin-2-one hydrochloride, pyrimidin-2-one hydrochloride
PubChem CID 122774
IUPAC Name 1H-pyrimidin-2-one;hydrochloride
SMILES C1=CNC(=O)N=C1.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC121860050
SDS View Documents
Thermo Scientific Chemicals
121860050
5 g Glass bottle
Each for $40.16
Only null left
 
AC121861000
SDS View Documents
Thermo Scientific Chemicals
121861000
100 g Glass bottle
Each for $389.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38353-09-2
132.55
IAJINJSFYTZPEJ-UHFFFAOYSA-N
122774
C1=CNC(=O)N=C1.Cl
C4H4N2O·ClH
MFCD00012781
2-hydroxypyrimidine hydrochloride, pyrimidin-2-ol hydrochloride, 2 1h-pyrimidinone, monohydrochloride, 2-hydroxypyrimidine hcl, 2-pyrimidinone hydrochloride, pyrimidin-2 1h-one hydrochloride, 2-hydroxypyrimidinehydrochloride, pyrimidin-2-one monohydrochloride, 1,2-dihydropyrimidin-2-one hydrochloride, pyrimidin-2-one hydrochloride
1H-pyrimidin-2-one;hydrochloride

Specifications

38353-09-2
100.0
Beige to Yellow
98%
C4H4N2O·ClH
5 g
2-hydroxypyrimidine hydrochloride, pyrimidin-2-ol hydrochloride, 2 1h-pyrimidinone, monohydrochloride, 2-hydroxypyrimidine hcl, 2-pyrimidinone hydrochloride, pyrimidin-2 1h-one hydrochloride, 2-hydroxypyrimidinehydrochloride, pyrimidin-2-one monohydrochloride, 1,2-dihydropyrimidin-2-one hydrochloride, pyrimidin-2-one hydrochloride
C1=CNC(=O)N=C1.Cl
132.55
132.55
Crystalline Solid
97.5
203°C to 205°C
Authentic
Glass bottle
MFCD00012781
24, I, 231
IAJINJSFYTZPEJ-UHFFFAOYSA-N
1H-pyrimidin-2-one;hydrochloride
122774
98%
2-Hydroxypyrimidine hydrochloride, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 253-897-8

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only