Learn More
2-Indanylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals
$164.00 - $428.68
Chemical Identifiers
CAS | 608534-44-7 |
---|---|
Molecular Formula | C15H21BO2 |
Molecular Weight (g/mol) | 244.14 |
MDL Number | MFCD03788726 |
InChI Key | YRTKSYXHFWUPAZ-UHFFFAOYSA-N |
Synonym | 2-indanylboronic acid pinacol ester, 2-2,3-dihydro-1h-inden-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-indanylboronicacidpinacolester, 2-indan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
PubChem CID | 24208876 |
IUPAC Name | 2-(2,3-dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | CC1(C)OB(OC1(C)C)C1CC2=CC=CC=C2C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL19535MD
|
Thermo Scientific Chemicals
L19535MD |
250 mg |
Each for $164.00
|
|
|||||
AAL1953503
|
Thermo Scientific Chemicals
L1953503 |
1 g |
Each for $428.68
|
|
|||||
Description
Boronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBoronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.
Solubility
Difficult to mix in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
608534-44-7 | |
244.14 | |
YRTKSYXHFWUPAZ-UHFFFAOYSA-N | |
24208876 | |
CC1(C)OB(OC1(C)C)C1CC2=CC=CC=C2C1 |
C15H21BO2 | |
MFCD03788726 | |
2-indanylboronic acid pinacol ester, 2-2,3-dihydro-1h-inden-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-indanylboronicacidpinacolester, 2-indan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
2-(2,3-dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
608534-44-7 | |
1.504 | |
250 mg | |
Difficult to mix in water. | |
CC1(C)OB(OC1(C)C)C1CC2=CC=CC=C2C1 | |
244.14 | |
244.14 | |
2-Indanylboronic acid pinacol ester |
C15H21BO2 | |
MFCD03788726 | |
2-indanylboronic acid pinacol ester, 2-2,3-dihydro-1h-inden-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-indanylboronicacidpinacolester, 2-indan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
YRTKSYXHFWUPAZ-UHFFFAOYSA-N | |
2-(2,3-dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
24208876 | |
95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only