missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Iodo-1,1':3',1″-terphenyl, 99%, Thermo Scientific Chemicals

$174.95 - $584.38

Chemical Identifiers

CAS 82777-09-1
Molecular Formula C18H13I
Molecular Weight (g/mol) 356.21
MDL Number MFCD00185026
InChI Key RLZYBGOJAWOQMK-UHFFFAOYSA-N
Synonym 2'-iodo-1,1':3',1-terphenyl, rlzybgojawoqmk-uhfffaoysa, 2'-iodo-1,1':3',1-terbenzene, 2-iodo-3-phenyl-1,1'-biphenyl, 1,1':3',1-terphenyl, 2'-iodo, 2'-iodo-1,1',3',1 terphenyl
PubChem CID 4078433
IUPAC Name 2-iodo-1,3-diphenylbenzene
SMILES IC1=C(C=CC=C1C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2670403
View Documents
Thermo Scientific Chemicals
H2670403
1 g
Each for $174.95
 
AAH2670406
View Documents
Thermo Scientific Chemicals
H2670406
5 g
Each for $584.38
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

82777-09-1
356.21
RLZYBGOJAWOQMK-UHFFFAOYSA-N
4078433
IC1=C(C=CC=C1C1=CC=CC=C1)C1=CC=CC=C1
C18H13I
MFCD00185026
2'-iodo-1,1':3',1-terphenyl, rlzybgojawoqmk-uhfffaoysa, 2'-iodo-1,1':3',1-terbenzene, 2-iodo-3-phenyl-1,1'-biphenyl, 1,1':3',1-terphenyl, 2'-iodo, 2'-iodo-1,1',3',1 terphenyl
2-iodo-1,3-diphenylbenzene

Specifications

82777-09-1
C18H13I
1 g
2'-iodo-1,1':3',1-terphenyl, rlzybgojawoqmk-uhfffaoysa, 2'-iodo-1,1':3',1-terbenzene, 2-iodo-3-phenyl-1,1'-biphenyl, 1,1':3',1-terphenyl, 2'-iodo, 2'-iodo-1,1',3',1 terphenyl
IC1=C(C=CC=C1C1=CC=CC=C1)C1=CC=CC=C1
356.21
356.21
2'-Iodo-1,1':3',1''-terphenyl
113°C to 116°C
MFCD00185026
Light sensitive
RLZYBGOJAWOQMK-UHFFFAOYSA-N
2-iodo-1,3-diphenylbenzene
4078433
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only