Learn More
2-Iodo-4-nitrotoluene, 97%, Thermo Scientific Chemicals
$78.30 - $248.85
Chemical Identifiers
CAS | 7745-92-8 |
---|---|
Molecular Formula | C7H6INO2 |
Molecular Weight (g/mol) | 263.03 |
MDL Number | MFCD00024329 |
InChI Key | BUQSRXQJUZTIEW-UHFFFAOYSA-N |
Synonym | 2-iodo-4-nitrotoluene, benzene, 2-iodo-1-methyl-4-nitro, 2-iodo-1-methyl-4-nitro-benzene, pubchem3957, acmc-209pa2, toluene, 2-iodo-4-nitro, timtec-bb sbb005821 |
PubChem CID | 82188 |
IUPAC Name | 2-iodo-1-methyl-4-nitrobenzene |
SMILES | CC1=CC=C(C=C1I)[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2149106
|
Thermo Scientific Chemicals
B2149106 |
5 g |
Each for $78.30
|
|
|||||
AAB2149114
|
Thermo Scientific Chemicals
B2149114 |
25 g |
Each for $248.85
|
|
|||||
Description
2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.
Solubility
Soluble in toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Light sensitive.
Chemical Identifiers
7745-92-8 | |
263.03 | |
BUQSRXQJUZTIEW-UHFFFAOYSA-N | |
82188 | |
CC1=CC=C(C=C1I)[N+]([O-])=O |
C7H6INO2 | |
MFCD00024329 | |
2-iodo-4-nitrotoluene, benzene, 2-iodo-1-methyl-4-nitro, 2-iodo-1-methyl-4-nitro-benzene, pubchem3957, acmc-209pa2, toluene, 2-iodo-4-nitro, timtec-bb sbb005821 | |
2-iodo-1-methyl-4-nitrobenzene |
Specifications
7745-92-8 | |
MFCD00024329 | |
Light sensitive | |
Soluble in toluene. | |
CC1=CC=C(C=C1I)[N+]([O-])=O | |
263.03 | |
263.03 | |
2-Iodo-4-nitrotoluene |
C7H6INO2 | |
5 g | |
2-iodo-4-nitrotoluene, benzene, 2-iodo-1-methyl-4-nitro, 2-iodo-1-methyl-4-nitro-benzene, pubchem3957, acmc-209pa2, toluene, 2-iodo-4-nitro, timtec-bb sbb005821 | |
BUQSRXQJUZTIEW-UHFFFAOYSA-N | |
2-iodo-1-methyl-4-nitrobenzene | |
82188 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 231-808-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only