Learn More
2-Iodo-p-xylene, 98+%, Thermo Scientific Chemicals
$177.75 - $718.19
Chemical Identifiers
CAS | 1122-42-5 |
---|---|
Molecular Formula | C8H9I |
Molecular Weight (g/mol) | 232.064 |
MDL Number | MFCD00013708 |
InChI Key | WYZVNUSNUCABRF-UHFFFAOYSA-N |
Synonym | 2-iodo-p-xylene, 1,4-dimethyl-2-iodobenzene, p-xylene, 2-iodo, 1-iodo-2,5-dimethylbenzene, benzene, 2-iodo-1,4-dimethyl, 2-iodo-4-xylene, 2,5-dimethyliodobenzene, 2-iodo-1,4-dimethyl-benzene, 2,5-dimethyl-1-iodobenzene, pubchem3875 |
PubChem CID | 70731 |
IUPAC Name | 2-iodo-1,4-dimethylbenzene |
SMILES | CC1=CC(=C(C=C1)C)I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0105509
|
Thermo Scientific Chemicals
L0105509 |
10 g |
Each for $177.75
|
|
|||||
AAL0105518
|
Thermo Scientific Chemicals
L0105518 |
50 g |
Each for $718.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1122-42-5 | |
232.064 | |
WYZVNUSNUCABRF-UHFFFAOYSA-N | |
70731 | |
CC1=CC(=C(C=C1)C)I |
C8H9I | |
MFCD00013708 | |
2-iodo-p-xylene, 1,4-dimethyl-2-iodobenzene, p-xylene, 2-iodo, 1-iodo-2,5-dimethylbenzene, benzene, 2-iodo-1,4-dimethyl, 2-iodo-4-xylene, 2,5-dimethyliodobenzene, 2-iodo-1,4-dimethyl-benzene, 2,5-dimethyl-1-iodobenzene, pubchem3875 | |
2-iodo-1,4-dimethylbenzene |
Specifications
1122-42-5 | |
229°C to 230°C | |
1.598 | |
MFCD00013708 | |
2-iodo-p-xylene, 1,4-dimethyl-2-iodobenzene, p-xylene, 2-iodo, 1-iodo-2,5-dimethylbenzene, benzene, 2-iodo-1,4-dimethyl, 2-iodo-4-xylene, 2,5-dimethyliodobenzene, 2-iodo-1,4-dimethyl-benzene, 2,5-dimethyl-1-iodobenzene, pubchem3875 | |
CC1=CC(=C(C=C1)C)I | |
232.064 | |
232.07 | |
2-Iodo-p-xylene |
1.617 | |
10 g | |
C8H9I | |
1905728 | |
WYZVNUSNUCABRF-UHFFFAOYSA-N | |
2-iodo-1,4-dimethylbenzene | |
70731 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 214-347-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only