missing translation for 'onlineSavingsMsg'
Learn More

2'-Iodoacetophenone, 98+%

CAS: 2142-70-3 | C8H7IO | 246.047 g/mol

$67.44 - $201.25

Chemical Identifiers

CAS 2142-70-3
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.047
MDL Number MFCD00094998
InChI Key XDXCBCXNCQGZPG-UHFFFAOYSA-N
Synonym 2'-iodoacetophenone, 1-2-iodophenyl ethanone, o-iodoacetophenone, 1-2-iodophenyl ethan-1-one, o-iodobenzoyl, acetophenone, o-iodo, acetophenone, 2'-iodo, ethanone, 1-iodophenyl, ethanone, 1-2-iodophenyl, 1-acetyl-2-iodobenzene
PubChem CID 240431
IUPAC Name 1-(2-iodophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2534503
View Documents
Thermo Scientific Chemicals
B2534503
1 g
Each for $67.44
Only null left
 
AAB2534506
View Documents
Thermo Scientific Chemicals
B2534506
5 g
Each for $201.25
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2142-70-3
246.047
XDXCBCXNCQGZPG-UHFFFAOYSA-N
240431
CC(=O)C1=CC=CC=C1I
C8H7IO
MFCD00094998
2'-iodoacetophenone, 1-2-iodophenyl ethanone, o-iodoacetophenone, 1-2-iodophenyl ethan-1-one, o-iodobenzoyl, acetophenone, o-iodo, acetophenone, 2'-iodo, ethanone, 1-iodophenyl, ethanone, 1-2-iodophenyl, 1-acetyl-2-iodobenzene
1-(2-iodophenyl)ethanone

Specifications

2142-70-3
139°C to 140°C (12 mmHg)
C8H7IO
MFCD00094998
2325078
2'-iodoacetophenone, 1-2-iodophenyl ethanone, o-iodoacetophenone, 1-2-iodophenyl ethan-1-one, o-iodobenzoyl, acetophenone, o-iodo, acetophenone, 2'-iodo, ethanone, 1-iodophenyl, ethanone, 1-2-iodophenyl, 1-acetyl-2-iodobenzene
CC(=O)C1=CC=CC=C1I
246.047
246.05
2'-Iodoacetophenone
1.746
103°C (218°F)
1.618
1 g
Light sensitive
XDXCBCXNCQGZPG-UHFFFAOYSA-N
1-(2-iodophenyl)ethanone
240431
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only