missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodoaniline, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 122301000

Catalog No. AC122301000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Iodoaniline
97.5
>110°C
98%
C6H6IN
100g
o-iodoaniline, benzenamine, 2-iodo, 2-iodo aniline, 2-iodobenzenamine, o-aminoiodobenzene, aniline, o-iodo, aniline, 2-iodo, benzenamine, iodo, 2-iodo-phenylamine, 2-iodophenylamine
UBPDKIDWEADHPP-UHFFFAOYSA-N
2-iodoaniline
11995
98%
615-43-0
100.0
Authentic
Glass bottle
MFCD00007680
12, 669
Solubility in water: slightly soluble
NC1=CC=CC=C1I
219.03
219.02
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 210-426-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only