missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodobutane, 99%, stab. with copper, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2061936

 View more versions of this product

Catalog No. AAB2061936


Explore available promotions

Description

Description

It is used as a solvent organic synthesis and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a solvent organic synthesis and as pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in alcohol and diethyl ether.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Specifications

2-Iodobutane
513-48-4
1.597
21°C (70°F)
C4H9I
CH3CH2CHICH3
500 g
1718777
14,1574
Insoluble in water. Soluble in alcohol and diethyl ether.
CCC(C)I
184.02
184.02
stabilized with copper
-104°C
119°C to 120°C
Ether-like
1.4991
MFCD00001072
UN2390
Light sensitive
sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide
IQRUSQUYPCHEKN-UHFFFAOYNA-N
2-iodobutane
10559
99%

Chemical Identifiers

513-48-4
184.02
IQRUSQUYPCHEKN-UHFFFAOYNA-N
10559
CCC(C)I
C4H9I
MFCD00001072
sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide
2-iodobutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-IODOBUTANE

EINECSNumber : 208-163-1

RTECSNumber : EK4410000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only