missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Isocyanatobenzonitrile, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals BTB13667DE
Specifications
2-Isocyanatobenzonitrile | |
95% | |
CNC6H4NCO | |
2-cyanophenyl isocyanate, benzonitrile,2-isocyanato, 2-isocyanato-benzonitrile, 2-isocyanatobenzenecarbonitrile, 2-cyanophenylisocyanate, benzonitrile, 2-isocyanato, acmc-1ap1b | |
C1=CC=C(C(=C1)C#N)N=C=O | |
144.133 | |
144.13 |
42066-86-4 | |
C8H4N2O | |
5g | |
FQLNVWAFXQUXGW-UHFFFAOYSA-N | |
2-isocyanatobenzonitrile | |
2735873 | |
95% |