missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Isocyanatoethyl Acrylate (stabilized with BHT) 98.0+%, TCI America™
$159.79 - $510.32
Chemical Identifiers
CAS | 13641-96-8 |
---|---|
Molecular Formula | C6H7NO3 |
Molecular Weight (g/mol) | 141.126 |
MDL Number | MFCD11112219 |
InChI Key | DPNXHTDWGGVXID-UHFFFAOYSA-N |
Synonym | Acrylic Acid 2-Isocyanatoethyl Ester, 2-(Acryloyloxy)ethyl Isocyanate |
PubChem CID | 3014768 |
IUPAC Name | 2-isocyanatoethyl prop-2-enoate |
SMILES | C=CC(=O)OCCN=C=O |
Chemical Identifiers
13641-96-8 | |
141.126 | |
DPNXHTDWGGVXID-UHFFFAOYSA-N | |
3014768 | |
C=CC(=O)OCCN=C=O |
C6H7NO3 | |
MFCD11112219 | |
Acrylic Acid 2-Isocyanatoethyl Ester, 2-(Acryloyloxy)ethyl Isocyanate | |
2-isocyanatoethyl prop-2-enoate |
Specifications
13641-96-8 | |
82°C | |
MFCD11112219 | |
2206 | |
DPNXHTDWGGVXID-UHFFFAOYSA-N | |
2-isocyanatoethyl prop-2-enoate | |
3014768 | |
≥98.0% (GC) | |
2-Isocyanatoethyl Acrylate (stabilized with BHT) |
Colorless | |
C6H7NO3 | |
5 g | |
Acrylic Acid 2-Isocyanatoethyl Ester, 2-(Acryloyloxy)ethyl Isocyanate | |
C=CC(=O)OCCN=C=O | |
141.126 | |
141.13 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-4063
TSCA : No
Recommended Storage : Refrigerator