missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Isopropoxyphenylboronic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 138008-97-6 |
---|---|
Molecular Formula | C9H13BO3 |
Molecular Weight (g/mol) | 180.01 |
InChI Key | XDMKBIIRBDPSOE-UHFFFAOYSA-N |
Synonym | 2-isopropoxyphenylboronic acid, 2-isopropoxyphenyl boronic acid, 2-isopoxybenzeneboronic acid, 2-isopropoxybenzeneboronic acid, boronic acid, 2-1-methylethoxy phenyl, 2-propan-2-yloxy phenylboronic acid, 2-isopropoxyphenylboronicacid, 2-propan-2-yloxyphenyl boronic acid, akos brn-0366, pubchem13297 |
PubChem CID | 5084099 |
IUPAC Name | (2-propan-2-yloxyphenyl)boronic acid |
SMILES | B(C1=CC=CC=C1OC(C)C)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC431380100
|
Thermo Scientific Chemicals
431380100 |
10g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
138008-97-6 | |
180.01 | |
2-isopropoxyphenylboronic acid, 2-isopropoxyphenyl boronic acid, 2-isopoxybenzeneboronic acid, 2-isopropoxybenzeneboronic acid, boronic acid, 2-1-methylethoxy phenyl, 2-propan-2-yloxy phenylboronic acid, 2-isopropoxyphenylboronicacid, 2-propan-2-yloxyphenyl boronic acid, akos brn-0366, pubchem13297 | |
(2-propan-2-yloxyphenyl)boronic acid |
C9H13BO3 | |
XDMKBIIRBDPSOE-UHFFFAOYSA-N | |
5084099 | |
B(C1=CC=CC=C1OC(C)C)(O)O |
Specifications
138008-97-6 | |
100.0 | |
110°C | |
96% min. (HPLC) | |
10g | |
XDMKBIIRBDPSOE-UHFFFAOYSA-N | |
(2-propan-2-yloxyphenyl)boronic acid | |
5084099 | |
97% |
96.0 | |
199.0°C | |
Authentic | |
C9H13BO3 | |
2-isopropoxyphenylboronic acid, 2-isopropoxyphenyl boronic acid, 2-isopoxybenzeneboronic acid, 2-isopropoxybenzeneboronic acid, boronic acid, 2-1-methylethoxy phenyl, 2-propan-2-yloxy phenylboronic acid, 2-isopropoxyphenylboronicacid, 2-propan-2-yloxyphenyl boronic acid, akos brn-0366, pubchem13297 | |
B(C1=CC=CC=C1OC(C)C)(O)O | |
180.01 | |
180.01 | |
2-Isopropoxyphenylboronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
GHS Signal Word: Warning
RUO â Research Use Only