missing translation for 'onlineSavingsMsg'
Learn More

2-Ketoglutaric acid, 98%

CAS: 328-50-7 | C5H6O5 | 146.098 g/mol

$65.67 - $531.36

Chemical Identifiers

CAS 328-50-7
Molecular Formula C5H6O5
Molecular Weight (g/mol) 146.098
MDL Number MFCD00004165
InChI Key KPGXRSRHYNQIFN-UHFFFAOYSA-N
Synonym 2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid
PubChem CID 51
ChEBI CHEBI:30915
IUPAC Name 2-oxopentanedioic acid
SMILES C(CC(=O)O)C(=O)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1025614
View Documents
Thermo Scientific Chemicals
A1025614
25 g
Each for $65.67
Only null left
 
AAA1025622
View Documents
Thermo Scientific Chemicals
A1025622
100 g
Each for $142.47
Only null left
 
AAA1025636
View Documents
Thermo Scientific Chemicals
A1025636
500 g
Each for $531.36
Only null left
 
Description

Description

2-Ketoglutaric acid is used as an intermediate in the kreb’s cycle catalyzed by glutamate dehydrogenase enzyme on glutamate. It is also used in dietary supplements to improve protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ketoglutaric acid is used as an intermediate in the kreb’s cycle catalyzed by glutamate dehydrogenase enzyme on glutamate. It is also used in dietary supplements to improve protein synthesis.

Solubility
Soluble in water, methanol and dimethyl sulfoxide.

Notes
Store away from heat store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

328-50-7
146.098
KPGXRSRHYNQIFN-UHFFFAOYSA-N
51
2-oxopentanedioic acid
C5H6O5
MFCD00004165
2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid
CHEBI:30915
C(CC(=O)O)C(=O)C(=O)O

Specifications

328-50-7
C5H6O5
25 g
14,5303
Soluble in water,methanol and dimethyl sulfoxide.
C(CC(=O)O)C(=O)C(=O)O
146.098
CHEBI:30915
98%
113°C to 117°C
MFCD00004165
1705689
2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid
KPGXRSRHYNQIFN-UHFFFAOYSA-N
2-oxopentanedioic acid
51
146.1
2-Ketoglutaric acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P280-P305+P351+P338-P310

H318

EINECSNumber : 206-330-3

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only