missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Mercaptobenzimidazole, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 125440050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Mercaptobenzimidazole | |
583-39-1 | |
100.0 | |
>250°C | |
98% | |
C7H6N2S | |
5g | |
15, 1084 | |
Solubility in water: 2.6g/L (23°C). Other solubilities: soluble in acetone,ethyl acetate and ethanol | |
C1=CC=C2C(=C1)NC(=S)N2 | |
150.2 | |
150.2 | |
0.3% max. (in NaOH) |
98% | |
97.5 | |
451°C | |
Authentic | |
Glass bottle | |
MFCD00466107 | |
24, 119 | |
2-mercaptobenzimidazole, 1h-benzoimidazole-2-thiol, 2-benzimidazolethiol, 1h-benzimidazole-2-thiol, 1h-benzo d imidazole-2-thiol, o-phenylenethiourea, antioxidant mb, mercaptobenzimidazole, 2-mercaptobenzoimidazole, antiegene mb | |
YHMYGUUIMTVXNW-UHFFFAOYSA-N | |
1,3-dihydrobenzimidazole-2-thione | |
707035 | |
98% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 209-502-6
RTECSNumber : DE1050000
TSCA : TSCA
RUO â Research Use Only