Learn More
2-Mercaptoethanol, 99%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125472500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Mercaptoethanol | |
-100.0°C | |
1.1100g/mL | |
73°C | |
98.5% min. (GC) | |
C2H6OS | |
HSCH2CH2OH | |
250 mL | |
01,643 | |
15,5936 | |
Solubility in water: soluble. Other solubilities: miscible with alcohol,ether,benzene,soluble in acetone,miscible with most common organic solvents | |
OCCS | |
78.13 | |
3.43 mPa.s (20°C) | |
78.13 | |
Pure |
60-24-2 | |
Colorless to Yellow | |
157.0°C to 158.0°C | |
Authentic | |
Glass bottle | |
1.4990 to 1.5020 | |
MFCD00004890 | |
01,47 | |
1.11 | |
2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan | |
DGVVWUTYPXICAM-UHFFFAOYSA-N | |
2-sulfanylethanol | |
1567 | |
CHEBI:41218 | |
99% | |
Liquid |
Chemical Identifiers
60-24-2 | |
78.13 | |
DGVVWUTYPXICAM-UHFFFAOYSA-N | |
1567 | |
OCCS |
C2H6OS | |
MFCD00004890 | |
2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan | |
CHEBI:41218 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause an allergic skin reaction.
May cause damage to organs through prolon
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 200-464-6
RUO – Research Use Only