missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Mercaptoethanol, For HPLC derivatization (14.3 M), ≥99.0% (GC), MilliporeSigma™ Supelco™
2-Mercaptoethanol is a thiol compound showing growth promoting properties on mouse lymphoma L1210 cells in vitro. It also enhances viability, blast transformation and antibody formation in lymphocyte cultures.
$66.54 - $268.50
Specifications
Purity | ≥99.0% (GC) |
---|---|
Boiling Point | 157°C |
Density | 1.114 g/mL (at 25°C) |
Flash Point | 74°C - Closed Cup |
Vapor Density | 2.69 (vs. air) |
Vapor Pressure | 1 mmHg (20°C) |
Quantity | 1 mL |
Specifications
≥99.0% (GC) | |
1.114 g/mL (at 25°C) | |
2.69 (vs. air) | |
1 mL |
157°C | |
74°C - Closed Cup | |
1 mmHg (20°C) |
Safety and Handling
RTECSNumber : KL5600000