missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mercaptoethanol (Reagent), Fisher Chemical

Supplier:  Thermo Fisher Scientific O3446I100

Catalog No. O3446I-100


Explore available promotions

Description

Description

Specifications

Specifications

Mercaptoethanol
60-24-2
Yellow
5.2
Amber Glass/Poison Pack
HSCH2CH2OH
100 mL
2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
OCCS
78.13
CHEBI:41218
98.0% min.
Liquid
2-Mercaptoethanol
-100°C
1.110g/cm³
157°C
C2H6OS
MFCD00004890
2966
DGVVWUTYPXICAM-UHFFFAOYSA-N
2-sulfanylethan-1-ol
1567
78.13
Reagent

Chemical Identifiers

60-24-2
78.13
DGVVWUTYPXICAM-UHFFFAOYSA-N
1567
2-sulfanylethan-1-ol
C2H6OS
MFCD00004890
2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
CHEBI:41218
OCCS
Videos
Safety and Handling

Safety and Handling

DOTInformation : Class 6.1: Poison

SDS Documents

Documents

Promotions

Promotions