missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mercaptopyridine-N-oxide, sodium salt, 40 w/w % aqueous solution, Thermo Scientific™

Specifications

CAS 3811-73-2
Molecular Formula C5H4NNaOS
Molecular Weight (g/mol) 149.15
MDL Number MFCD01941547
InChI Key XNRNJIIJLOFJEK-UHFFFAOYSA-N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC257740010
View Documents
Thermo Scientific Chemicals
257740010
1L Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC257740025
View Documents
Thermo Scientific Chemicals
257740025
2.5L Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC257740050
View Documents
Thermo Scientific Chemicals
257740050
5L Plastic drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3811-73-2
149.15
XNRNJIIJLOFJEK-UHFFFAOYSA-N
19658
C1=CC(=S)N(C=C1)[O-].[Na+]
C5H4NNaOS
MFCD01941547
sodium omadine, sodium pyrithione, sodium 2-pyridylthio-n-oxide, pyrithione sodium salt, thione reagent, 2-pyridinethiol-1-oxide sodium salt, 2-mercaptopyridine n-oxide sodium salt, omadine sodium, sodium-2-pyridinethiol-1-oxide, sodium omadine van
sodium;1-oxidopyridine-2-thione
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only