missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methacryloyloxy-2-methyladamantane (stabilized with MEHQ) 97.0+%, TCI America™
$89.59 - $358.35
Chemical Identifiers
CAS | 177080-67-0 |
---|---|
Molecular Formula | C15H22O2 |
Molecular Weight (g/mol) | 234.339 |
MDL Number | MFCD04114354 |
InChI Key | FDYDISGSYGFRJM-UHFFFAOYSA-N |
Synonym | 2-Methyl-2-adamantyl Methacrylate, Methacrylic Acid 2-Methyl-2-adamantyl Ester |
PubChem CID | 9837566 |
IUPAC Name | (2-methyl-2-adamantyl) 2-methylprop-2-enoate |
SMILES | CC(=C)C(=O)OC1(C2CC3CC(C2)CC1C3)C |
Chemical Identifiers
177080-67-0 | |
234.339 | |
FDYDISGSYGFRJM-UHFFFAOYSA-N | |
9837566 | |
CC(=C)C(=O)OC1(C2CC3CC(C2)CC1C3)C |
C15H22O2 | |
MFCD04114354 | |
2-Methyl-2-adamantyl Methacrylate, Methacrylic Acid 2-Methyl-2-adamantyl Ester | |
(2-methyl-2-adamantyl) 2-methylprop-2-enoate |
Specifications
177080-67-0 | |
C15H22O2 | |
1 g | |
FDYDISGSYGFRJM-UHFFFAOYSA-N | |
(2-methyl-2-adamantyl) 2-methylprop-2-enoate | |
9837566 | |
≥97.0% (GC) | |
2-Methacryloyloxy-2-methyladamantane (stabilized with MEHQ) |
Yellow | |
MFCD04114354 | |
2-Methyl-2-adamantyl Methacrylate, Methacrylic Acid 2-Methyl-2-adamantyl Ester | |
CC(=C)C(=O)OC1(C2CC3CC(C2)CC1C3)C | |
234.339 | |
234.34 | |
Liquid |
Safety and Handling
EINECSNumber : (4)-1884
TSCA : No
Recommended Storage : Freezer