missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-3-methylpyrazine, 99%, Thermo Scientific Chemicals

$108.29 - $337.85

Chemical Identifiers

CAS 2847-30-5
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD00006127
InChI Key VKJIAEQRKBQLLA-UHFFFAOYSA-N
Synonym pyrazine, 2-methoxy-3-methyl, 2-methyl-3-methoxypyrazine, 2-methoxy-3-methyl-pyrazine, 2-methoxy-3-methyl pyrazine, unii-04o7cn9q85, pyrazine, 2-methoxy-3 or 5-methyl, pyrazine, 2-methyl-3-methoxy, almond pyrazine, 2-methoxy-3 or5-methylpyrazine, chocolate pyrazine a
PubChem CID 17898
IUPAC Name 2-methoxy-3-methylpyrazine
SMILES CC1=NC=CN=C1OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0806706
View Documents
Thermo Scientific Chemicals
L0806706
5 g
Each for $108.29
 
AAL0806714
View Documents
Thermo Scientific Chemicals
L0806714
25 g
Each for $337.85
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2847-30-5
124.143
VKJIAEQRKBQLLA-UHFFFAOYSA-N
17898
CC1=NC=CN=C1OC
C6H8N2O
MFCD00006127
pyrazine, 2-methoxy-3-methyl, 2-methyl-3-methoxypyrazine, 2-methoxy-3-methyl-pyrazine, 2-methoxy-3-methyl pyrazine, unii-04o7cn9q85, pyrazine, 2-methoxy-3 or 5-methyl, pyrazine, 2-methyl-3-methoxy, almond pyrazine, 2-methoxy-3 or5-methylpyrazine, chocolate pyrazine a
2-methoxy-3-methylpyrazine

Specifications

2847-30-5
159°C
C6H8N2O
MFCD00006127
UN1993
pyrazine, 2-methoxy-3-methyl, 2-methyl-3-methoxypyrazine, 2-methoxy-3-methyl-pyrazine, 2-methoxy-3-methyl pyrazine, unii-04o7cn9q85, pyrazine, 2-methoxy-3 or 5-methyl, pyrazine, 2-methyl-3-methoxy, almond pyrazine, 2-methoxy-3 or5-methylpyrazine, chocolate pyrazine a
CC1=NC=CN=C1OC
124.143
124.14
2-Methoxy-3-methylpyrazine
1.07
55°C (131°F)
1.5065
5 g
878809
VKJIAEQRKBQLLA-UHFFFAOYSA-N
2-methoxy-3-methylpyrazine
17898
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226-H315-H319
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c

H226-H302

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 220-651-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only