Learn More
2-Methoxy-4-nitroaniline, 99%
CAS: 97-52-9 | C7H8N2O3 | 168.15 g/mol
Supplier: Thermo Scientific Chemicals B2422036
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Methoxy-4-nitroaniline | |
| 138°C to 142°C | |
| C7H8N2O3 | |
| 500 g | |
| fast red b base, benzenamine, 2-methoxy-4-nitro, 4-nitro-o-anisidine, fast red base b, fast red b, red b base, diazo fast red b, 2-amino-5-nitroanisole, azoamine pink o, fast red 5na base | |
| COC1=CC(=CC=C1N)[N+]([O-])=O | |
| 168.15 | |
| 168.15 |
| 97-52-9 | |
| 1.211 | |
| MFCD00007363 | |
| 879619 | |
| GVBHRNIWBGTNQA-UHFFFAOYSA-N | |
| 2-methoxy-4-nitroaniline | |
| 7337 | |
| 99% |
Chemical Identifiers
| 97-52-9 | |
| 168.15 | |
| GVBHRNIWBGTNQA-UHFFFAOYSA-N | |
| 7337 | |
| COC1=CC(=CC=C1N)[N+]([O-])=O |
| C7H8N2O3 | |
| MFCD00007363 | |
| fast red b base, benzenamine, 2-methoxy-4-nitro, 4-nitro-o-anisidine, fast red base b, fast red b, red b base, diazo fast red b, 2-amino-5-nitroanisole, azoamine pink o, fast red 5na base | |
| 2-methoxy-4-nitroaniline |
Safety and Handling
GHS H Statement
H301-H311-H341-H332
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing genetic defects.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
EINECSNumber : 202-588-6
RTECSNumber : BZ7170000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only