missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-4-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2552303

 View more versions of this product

Catalog No. AAB2552303


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methoxy-4-nitrobenzoic acid
145°C to 148°C
MFCD00216573
2-methoxy-4-nitrobenzoic acid, 4-nitro-o-anisic acid, 4-nitro-2-methoxybenzoic acid, rarechem al bo 1464, 2-carboxy-5-nitroanisole, 2-methoxy-4-nitro-benzoic acid, benzoic acid, 2-methoxy-4-nitro, 4-nitro-2-methoxybenzoicacid, pubchem4977, acmc-20a5eb
COC1=CC(=CC=C1C(O)=O)[N+]([O-])=O
197.15
197.15
2597-56-0
C8H7NO5
1 g
KPJXEWJRJKEOCD-UHFFFAOYSA-N
2-methoxy-4-nitrobenzoic acid
75776
98%

Chemical Identifiers

2597-56-0
197.15
KPJXEWJRJKEOCD-UHFFFAOYSA-N
75776
COC1=CC(=CC=C1C(O)=O)[N+]([O-])=O
C8H7NO5
MFCD00216573
2-methoxy-4-nitrobenzoic acid, 4-nitro-o-anisic acid, 4-nitro-2-methoxybenzoic acid, rarechem al bo 1464, 2-carboxy-5-nitroanisole, 2-methoxy-4-nitro-benzoic acid, benzoic acid, 2-methoxy-4-nitro, 4-nitro-2-methoxybenzoicacid, pubchem4977, acmc-20a5eb
2-methoxy-4-nitrobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 219-998-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only