missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxycarbonylphenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 374538-03-1
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.97
MDL Number MFCD02179452
InChI Key ODAXNYMENLFYMY-UHFFFAOYSA-N
Synonym 2-methoxycarbonyl phenylboronic acid, 2-methoxycarbonyl benzeneboronic acid, 2-methoxycarbonyl phenyl boronic acid, 2-methoxycarbonylphenyl boronic acid, methyl 2-boronobenzoate, methyl 2-dihydroxyboranyl benzoate, o-methoxycarbonyl phenylboronic acid, 2-carbomethoxybenzeneboronic acid, 2-methoxycarbonylphenylboronicacid
PubChem CID 2773496
IUPAC Name (2-methoxycarbonylphenyl)boronic acid
SMILES B(C1=CC=CC=C1C(=O)OC)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC433910010
SDS View Documents
Thermo Scientific Chemicals
433910010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC433910050
SDS View Documents
Thermo Scientific Chemicals
433910050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

374538-03-1
179.97
ODAXNYMENLFYMY-UHFFFAOYSA-N
2773496
B(C1=CC=CC=C1C(=O)OC)(O)O
C8H9BO4
MFCD02179452
2-methoxycarbonyl phenylboronic acid, 2-methoxycarbonyl benzeneboronic acid, 2-methoxycarbonyl phenyl boronic acid, 2-methoxycarbonylphenyl boronic acid, methyl 2-boronobenzoate, methyl 2-dihydroxyboranyl benzoate, o-methoxycarbonyl phenylboronic acid, 2-carbomethoxybenzeneboronic acid, 2-methoxycarbonylphenylboronicacid
(2-methoxycarbonylphenyl)boronic acid

Specifications

374538-03-1
100.0
96% min. (HPLC)
C8H9BO4
1g
ODAXNYMENLFYMY-UHFFFAOYSA-N
(2-methoxycarbonylphenyl)boronic acid
2773496
97%
96.0
Authentic
Glass bottle
MFCD02179452
2-methoxycarbonyl phenylboronic acid, 2-methoxycarbonyl benzeneboronic acid, 2-methoxycarbonyl phenyl boronic acid, 2-methoxycarbonylphenyl boronic acid, methyl 2-boronobenzoate, methyl 2-dihydroxyboranyl benzoate, o-methoxycarbonyl phenylboronic acid, 2-carbomethoxybenzeneboronic acid, 2-methoxycarbonylphenylboronicacid
B(C1=CC=CC=C1C(=O)OC)(O)O
179.97
179.97
2-Methoxycarbonylphenylboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only