missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyfuran, 97%, Thermo Scientific Chemicals

$171.26 - $558.22

Chemical Identifiers

CAS 25414-22-6
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00003227
InChI Key OXCGHDNCMSOEBZ-UHFFFAOYSA-N
PubChem CID 117476
IUPAC Name 2-methoxyfuran
SMILES COC1=CC=CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1559303
View Documents
Thermo Scientific Chemicals
A1559303
1 g
Each for $171.26
 
AAA1559306
View Documents
Thermo Scientific Chemicals
A1559306
5 g
Each for $558.22
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25414-22-6
98.101
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
2-methoxyfuran
C5H6O2
MFCD00003227
117476
COC1=CC=CO1

Specifications

25414-22-6
110°C to 111°C
C5H6O2
MFCD00003227
UN3271
furan, 2-methoxy, 2-methoxy furan, unii-25y8i1p35t, 2-furyl methyl ether, furan,2-methoxy, pubchem19998, acmc-209gjs, 2-methoxyfuran, 2-furyl methyl ether #
COC1=CC=CO1
98.101
98.1
2-Methoxyfuran
1.065
10°C (50°F)
1.447
1 g
106270
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
2-methoxyfuran
117476
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c

H225-H301+H311+H331

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 246-953-8

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only