missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methoxynicotinic acid, 98%, Thermo Scientific Chemicals
$33.62 - $112.48
Chemical Identifiers
CAS | 16498-81-0 |
---|---|
Molecular Formula | C7H6NO3 |
Molecular Weight (g/mol) | 152.13 |
MDL Number | MFCD00085110 |
InChI Key | FTEZJSXSARPZHJ-UHFFFAOYSA-M |
Synonym | 2-methoxynicotinic acid, 2-methoxy-nicotinic acid, 2-methoxy-3-pyridinecarboxylic acid, 2-methoxy nicotinic acid, 3-pyridinecarboxylic acid, 2-methoxy, t6nj bo1 cvq wwn, maybridge1_004722, 2-methoxynicotinicacid, pubchem20354, acmc-1btxh |
PubChem CID | 708093 |
IUPAC Name | 2-methoxypyridine-3-carboxylic acid |
SMILES | COC1=NC=CC=C1C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1319303
|
Thermo Scientific Chemicals
A1319303 |
1 g |
Each for $33.62
|
|
||||||
AAA1319306
|
Thermo Scientific Chemicals
A1319306 |
5 g |
Each for $112.48
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16498-81-0 | |
152.13 | |
FTEZJSXSARPZHJ-UHFFFAOYSA-M | |
708093 | |
COC1=NC=CC=C1C([O-])=O |
C7H6NO3 | |
MFCD00085110 | |
2-methoxynicotinic acid, 2-methoxy-nicotinic acid, 2-methoxy-3-pyridinecarboxylic acid, 2-methoxy nicotinic acid, 3-pyridinecarboxylic acid, 2-methoxy, t6nj bo1 cvq wwn, maybridge1_004722, 2-methoxynicotinicacid, pubchem20354, acmc-1btxh | |
2-methoxypyridine-3-carboxylic acid |
Specifications
16498-81-0 | |
C7H6NO3 | |
1 g | |
FTEZJSXSARPZHJ-UHFFFAOYSA-M | |
2-methoxypyridine-3-carboxylic acid | |
708093 | |
98% |
147°C to 151°C | |
MFCD00085110 | |
2-methoxynicotinic acid, 2-methoxy-nicotinic acid, 2-methoxy-3-pyridinecarboxylic acid, 2-methoxy nicotinic acid, 3-pyridinecarboxylic acid, 2-methoxy, t6nj bo1 cvq wwn, maybridge1_004722, 2-methoxynicotinicacid, pubchem20354, acmc-1btxh | |
COC1=NC=CC=C1C([O-])=O | |
152.13 | |
153.14 | |
2-Methoxynicotinic acid |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only