missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methoxyphenyl Acetate 97.0+%, TCI America™
Supplier: TCI America A138110G
Specifications
2-Methoxyphenyl Acetate | |
Colorless | |
C9H10O3 | |
10 g | |
BHJHPYFAYGAPLS-UHFFFAOYSA-N | |
(2-methoxyphenyl) acetate | |
61155 | |
166.18 | |
Liquid |
613-70-7 | |
240°C | |
MFCD00017221 | |
Acetic Acid 2-Methoxyphenyl Ester, 2-Guaiacol Acetate, Acetic Acid 2-Guaiacol Ester, O-Acetylguaiacol | |
CC(=O)OC1=CC=CC=C1OC | |
166.176 | |
CHEBI:86645 | |
≥97.0% (GC) |
Chemical Identifiers
613-70-7 | |
166.176 | |
BHJHPYFAYGAPLS-UHFFFAOYSA-N | |
61155 | |
(2-methoxyphenyl) acetate |
C9H10O3 | |
MFCD00017221 | |
Acetic Acid 2-Methoxyphenyl Ester, 2-Guaiacol Acetate, Acetic Acid 2-Guaiacol Ester, O-Acetylguaiacol | |
CHEBI:86645 | |
CC(=O)OC1=CC=CC=C1OC |
Safety and Handling
EINECSNumber : (3)-0645
TSCA : Yes