missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

$120.93 - $400.61

Chemical Identifiers

CAS 93-25-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004321
InChI Key IVEWTCACRDEAOB-UHFFFAOYSA-N
Synonym 2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
PubChem CID 7134
IUPAC Name 2-(2-methoxyphenyl)acetic acid
SMILES COC1=CC=CC=C1CC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC177600250
SDS View Documents
Thermo Scientific Chemicals
177600250
25 g Glass bottle
Each for $120.93
 
AC177601000
SDS View Documents
Thermo Scientific Chemicals
177601000
100 g Glass bottle
Each for $400.61
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

93-25-4
166.18
IVEWTCACRDEAOB-UHFFFAOYSA-N
7134
COC1=CC=CC=C1CC(O)=O
C9H10O3
MFCD00004321
2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
2-(2-methoxyphenyl)acetic acid

Specifications

93-25-4
100.0
Cream-Pink to White
99%
C9H10O3
MFCD00004321
10, 188
Solubility in water: soluble. Other solubilities: soluble in ethenol,ethyl ether,acetone
COC1=CC=CC=C1CC(O)=O
166.18
166.18
Crystalline Powder
98.5
122.0°C to 125.0°C
Authentic
Glass bottle
CH3OC6H4CH2CO2H
25 g
2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
IVEWTCACRDEAOB-UHFFFAOYSA-N
2-(2-methoxyphenyl)acetic acid
7134
99%
2-Methoxyphenylacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning

EINECSNumber : 202-231-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only