missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylacetonitrile 98.0+%, TCI America™

$50.33 - $50.33

Chemical Identifiers

CAS 7035-03-2
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00001902
InChI Key DWJKILXTMUGXOU-UHFFFAOYSA-N
Synonym 2-methoxyphenyl acetonitrile, 2-methoxyphenylacetonitrile, 2-methoxybenzyl cyanide, 2-2-methoxyphenyl acetonitrile, benzeneacetonitrile, 2-methoxy, o-methoxyphenylacetonitrile, o-methoxybenzyl cyanide, o-methoxyphenyl acetonitrile, 2-2-methoxyphenyl ethanenitrile, 2-methoxybenzylcyanid
PubChem CID 81496
IUPAC Name 2-(2-methoxyphenyl)acetonitrile
SMILES COC1=CC=CC=C1CC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M085525G
View Documents
TCI America
M085525G
25 g
Each for $50.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7035-03-2
147.18
DWJKILXTMUGXOU-UHFFFAOYSA-N
81496
COC1=CC=CC=C1CC#N
C9H9NO
MFCD00001902
2-methoxyphenyl acetonitrile, 2-methoxyphenylacetonitrile, 2-methoxybenzyl cyanide, 2-2-methoxyphenyl acetonitrile, benzeneacetonitrile, 2-methoxy, o-methoxyphenylacetonitrile, o-methoxybenzyl cyanide, o-methoxyphenyl acetonitrile, 2-2-methoxyphenyl ethanenitrile, 2-methoxybenzylcyanid
2-(2-methoxyphenyl)acetonitrile

Specifications

7035-03-2
White-Yellow
C9H9NO
25 g
DWJKILXTMUGXOU-UHFFFAOYSA-N
2-(2-methoxyphenyl)acetonitrile
81496
≥98.0% (GC)
2-Methoxyphenylacetonitrile
68°C
97°C
MFCD00001902
2-methoxyphenyl acetonitrile, 2-methoxyphenylacetonitrile, 2-methoxybenzyl cyanide, 2-2-methoxyphenyl acetonitrile, benzeneacetonitrile, 2-methoxy, o-methoxyphenylacetonitrile, o-methoxybenzyl cyanide, o-methoxyphenyl acetonitrile, 2-2-methoxyphenyl ethanenitrile, 2-methoxybenzylcyanid
COC1=CC=CC=C1CC#N
147.18
147.18
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'