missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxythiophene 98.0+%, TCI America™

$180.81 - $615.14

Chemical Identifiers

CAS 16839-97-7
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00014526
InChI Key OKEHURCMYKPVFW-UHFFFAOYSA-N
Synonym thiophene, 2-methoxy, 2-methoxy thiophene, 2-methoxy-thiophene, 2-methyloxy thiophene, pubchem12545, acmc-1bwzz, 2-methoxythiophene, ksc183i2f
PubChem CID 85610
IUPAC Name 2-methoxythiophene
SMILES COC1=CC=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M10305G
View Documents
TCI America
M10305G
5 g
Each for $180.81
Only null left
 
M103025G
View Documents
TCI America
M103025G
25 g
Each for $615.14
Only null left
 
Description

Description

Specifications

Chemical Identifiers

16839-97-7
114.16
OKEHURCMYKPVFW-UHFFFAOYSA-N
85610
COC1=CC=CS1
C5H6OS
MFCD00014526
thiophene, 2-methoxy, 2-methoxy thiophene, 2-methoxy-thiophene, 2-methyloxy thiophene, pubchem12545, acmc-1bwzz, 2-methoxythiophene, ksc183i2f
2-methoxythiophene

Specifications

16839-97-7
154°C
MFCD00014526
1993
OKEHURCMYKPVFW-UHFFFAOYSA-N
2-methoxythiophene
85610
≥98.0% (GC)
2-Methoxythiophene
Yellow
C5H6OS
5 g
thiophene, 2-methoxy, 2-methoxy thiophene, 2-methoxy-thiophene, 2-methyloxy thiophene, pubchem12545, acmc-1bwzz, 2-methoxythiophene, ksc183i2f
COC1=CC=CS1
114.16
114.16
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'