Learn More
2-Methyl-1,4-naphthoquinone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 127181000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methyl-1, 4-naphthoquinone | |
58-27-5 | |
100.0 | |
Green-Yellow to Yellow | |
97.5% min. (HPLC) | |
C11H8O2 | |
100 g | |
15, 5899 | |
Solubility in water: insoluble in water. Other solubilities: soluble in alcohol (1g/60 mL),benzene (1g/10 mL,soluble in vegetable oils (1g/50 mL),moderately soluble in chloroform and carbon tetrac | |
CC1=CC(=O)C2=CC=CC=C2C1=O | |
172.18 | |
CHEBI:28869 | |
98% |
98% | |
97.5 | |
105.0°C to 108.0°C | |
Authentic | |
Glass bottle | |
MFCD00001681 | |
07, II, 656 | |
menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine | |
MJVAVZPDRWSRRC-UHFFFAOYSA-N | |
2-methylnaphthalene-1,4-dione | |
4055 | |
172.18 | |
Powder |
Chemical Identifiers
58-27-5 | |
172.18 | |
MJVAVZPDRWSRRC-UHFFFAOYSA-N | |
4055 | |
2-methylnaphthalene-1,4-dione |
C11H8O2 | |
MFCD00001681 | |
menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine | |
CHEBI:28869 | |
CC1=CC(=O)C2=CC=CC=C2C1=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 200-372-6
RTECSNumber : QL9100000
TSCA : TSCA
RUO – Research Use Only