missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1-phenyl-1-propanol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2072914

 View more versions of this product

Catalog No. AAB2072914


Explore available promotions

Description

Description

It is used in the large-scale preparation of (R)-and (S)-1-(2, 4, 6-triisopropylphenyl) ethanol and also in the baker's yeast mediated asymmetric reduction of cinnamaldehyde derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the large-scale preparation of (R)-and (S)-1-(2, 4, 6-triisopropylphenyl) ethanol and also in the baker′s yeast mediated asymmetric reduction of cinnamaldehyde derivatives.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

2-Methyl-1-phenyl-1-propanol
0.964
86°C (187°F)
1.513
25 g
Not miscible or difficult to mix in water.
CC(C)C(C1=CC=CC=C1)O
150.221
150.22
611-69-8
124°C to 125°C (15 mmHg)
C10H14O
MFCD00065000
2-methyl-1-phenyl-1-propanol, .alpha.-isopropylbenzyl alcohol, isopropyl benzyl alcohol, 2-methyl-1-phenylpropanol, benzenemethanol, .alpha.-1-methylethyl, 1-phenyl-2-methylpropyl alcohol, 1r-2-methyl-1-phenyl-propan-1-ol, phenylisopropylcarbinol, isopropylphenylcarbinol, acmc-20anr9
GMDYDZMQHRTHJA-UHFFFAOYSA-N
2-methyl-1-phenylpropan-1-ol
95626
98%

Chemical Identifiers

611-69-8
150.221
GMDYDZMQHRTHJA-UHFFFAOYSA-N
95626
CC(C)C(C1=CC=CC=C1)O
C10H14O
MFCD00065000
2-methyl-1-phenyl-1-propanol, .alpha.-isopropylbenzyl alcohol, isopropyl benzyl alcohol, 2-methyl-1-phenylpropanol, benzenemethanol, .alpha.-1-methylethyl, 1-phenyl-2-methylpropyl alcohol, 1r-2-methyl-1-phenyl-propan-1-ol, phenylisopropylcarbinol, isopropylphenylcarbinol, acmc-20anr9
2-methyl-1-phenylpropan-1-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 210-274-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only