missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-butanol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A183040D

 View more versions of this product

Catalog No. AAA183040D

Explore available promotions

Description

Description

2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.

Solubility
Miscible with alcohol, ether, benzene, chloroform, glycerol, oils and acetone.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

2-Methyl-2-butanol
∼-12°C
101°C to 103°C
0.98
C5H12O
MFCD00004478
UN1105
14,7140
Miscible with alcohol,ether,benzene,chloroform,glycerol,oils and acetone.
CCC(C)(C)O
88.15
88.15
Liquid
75-85-4
0.805
20°C (68°F)
Unpleasant
1.405
10,000 mL
1361351
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
MSXVEPNJUHWQHW-UHFFFAOYSA-N
2-methylbutan-2-ol
6405
98%

Chemical Identifiers

75-85-4
88.15
MSXVEPNJUHWQHW-UHFFFAOYSA-N
6405
CCC(C)(C)O
C5H12O
MFCD00004478
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
2-methylbutan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H332-H315-H335
Highly flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c

H225-H312+H332-H315-H318-H335-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANOLS

EINECSNumber : 200-908-9

RTECSNumber : SC0175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only