missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-propanethiol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 149020500

 View more versions of this product

Catalog No. AC149020500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methyl-2-propanethiol
75-66-1
100.0
Colorless
62°C to 65°C
Authentic
Glass bottle
1.4220 to 1.424
MFCD00004857
01, 383
15, 1582
Solubility in water: slightly soluble. Other solubilities: soluble in liquid h2s,very soluble in alcohol and ether
CC(C)(C)S
90.18
0.638 mPa.s (20°C)
99%
99%
98.5
-1.1°C
0.8000g/mL
-24°C
98.5% min. (GC)
C4H10S
(CH3)3CSH
50 mL
0.8
2-methyl-2-propanethiol, tert-butyl mercaptan, tert-butylthiol, tert-butanethiol, 2-propanethiol, 2-methyl, tert-butylmercaptan, t-butyl mercaptan, t-butylmercaptan, 2-isobutanethiol, tertiary-butyl mercaptan
WMXCDAVJEZZYLT-UHFFFAOYSA-N
2-methylpropane-2-thiol
6387
90.18
Liquid

Chemical Identifiers

75-66-1
90.18
WMXCDAVJEZZYLT-UHFFFAOYSA-N
6387
CC(C)(C)S
C4H10S
MFCD00004857
2-methyl-2-propanethiol, tert-butyl mercaptan, tert-butylthiol, tert-butanethiol, 2-propanethiol, 2-methyl, tert-butylmercaptan, t-butyl mercaptan, t-butylmercaptan, 2-isobutanethiol, tertiary-butyl mercaptan
2-methylpropane-2-thiol
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Ground/Bond container and receiving equipment.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 200-890-2

RTECSNumber : TZ7660000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only