missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3,5-dinitrobenzoic acid, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1455522

 View more versions of this product

Catalog No. AAA1455522


Explore available promotions

Description

Description

2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

2-Methyl-3,5-dinitrobenzoic acid
203°C to 207°C
MFCD00007161
2622906
Soluble in water.
CC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O
226.14
226.15
28169-46-2
C8H6N2O6
100 g
2-methyl-3,5-dinitrobenzoic acid, 3,5-dinitro-2-methylbenzoic acid, 3,5-dinitro-o-toluic acid, benzoic acid, 2-methyl-3,5-dinitro, 3,5-dinitrotoluic acid, o-toluic acid, 3,5-dinitro, 2-methyl-3,5-dinitro-benzoic acid, 3,5-dinitro-2-methyl-benzoic acid, acmc-1cfeu, o-toluic acid,5-dinitro
CDVNZMKTJIBBBV-UHFFFAOYSA-N
2-methyl-3,5-dinitrobenzoic acid
98824
98%

Chemical Identifiers

28169-46-2
226.14
CDVNZMKTJIBBBV-UHFFFAOYSA-N
98824
CC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O
C8H6N2O6
MFCD00007161
2-methyl-3,5-dinitrobenzoic acid, 3,5-dinitro-2-methylbenzoic acid, 3,5-dinitro-o-toluic acid, benzoic acid, 2-methyl-3,5-dinitro, 3,5-dinitrotoluic acid, o-toluic acid, 3,5-dinitro, 2-methyl-3,5-dinitro-benzoic acid, 3,5-dinitro-2-methyl-benzoic acid, acmc-1cfeu, o-toluic acid,5-dinitro
2-methyl-3,5-dinitrobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 248-880-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only