missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-buten-2-ol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 159672500

 View more versions of this product

Catalog No. AC159672500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methyl-3-buten-2-ol
115-18-4
100.0
Colorless to Yellow
98.0°C to 99.0°C
Authentic
Glass bottle
1.4160 to 1.4180
MFCD00004470
01, 444
2-methyl-3-buten-2-ol, 1,1-dimethylallyl alcohol, methylbutenol, 3-buten-2-ol, 2-methyl, 3-hydroxy-3-methyl-1-butene, dimethylvinylcarbinol, dimethylvinylmethanol, vinyldimethylcarbinol, isoprenyl alcohol, 3-methyl-1-buten-3-ol
HNVRRHSXBLFLIG-UHFFFAOYSA-N
2-methylbut-3-en-2-ol
8257
97%
97%
96.0
-43.0°C
0.8200g/mL
13°C
96% min. (GC)
C5H10O
H2C=CHC(CH3)2OH
250 mL
0.82
Solubility in water: 100g/L (20 c)
CC(C)(C=C)O
86.13
86.13
Liquid

Chemical Identifiers

115-18-4
86.13
HNVRRHSXBLFLIG-UHFFFAOYSA-N
8257
CC(C)(C=C)O
C5H10O
MFCD00004470
2-methyl-3-buten-2-ol, 1,1-dimethylallyl alcohol, methylbutenol, 3-buten-2-ol, 2-methyl, 3-hydroxy-3-methyl-1-butene, dimethylvinylcarbinol, dimethylvinylmethanol, vinyldimethylcarbinol, isoprenyl alcohol, 3-methyl-1-buten-3-ol
2-methylbut-3-en-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-068-4

RTECSNumber : EM9472000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only