missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-furoic acid, 98%, Thermo Scientific Chemicals

$109.21 - $325.05

Chemical Identifiers

CAS 6947-94-0
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00092314
InChI Key CFGQZVOVFIZRMN-UHFFFAOYSA-N
PubChem CID 244756
IUPAC Name 2-methylfuran-3-carboxylic acid
SMILES CC1=C(C=CO1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2492806
View Documents
Thermo Scientific Chemicals
B2492806
5 g
Each for $109.21
 
AAB2492814
View Documents
Thermo Scientific Chemicals
B2492814
25 g
Each for $325.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6947-94-0
126.111
CFGQZVOVFIZRMN-UHFFFAOYSA-N
2-methylfuran-3-carboxylic acid
C6H6O3
MFCD00092314
244756
CC1=C(C=CO1)C(=O)O

Specifications

6947-94-0
C6H6O3
5 g
2-methyl-3-furoic acid, 2-methyl-3-furancarboxylic acid, 3-furancarboxylic acid, 2-methyl, 2-methyl-furan-3-carboxylic acid, 3-furancarboxylic acid,2-methyl, zlchem 1156, acmc-1b2rk, 2-methyl-3furan carboxylic acid, 2-methyl-3-furan carboxylic acid
CC1=C(C=CO1)C(=O)O
126.111
126.11
2-Methyl-3-furoic acid
97°C to 99°C
MFCD00092314
112229
CFGQZVOVFIZRMN-UHFFFAOYSA-N
2-methylfuran-3-carboxylic acid
244756
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only