missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-nitrobenzyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 187400010

Catalog No. AC187400010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-Methyl-3-nitrobenzyl chloride
60468-54-4
100.0
96% min. (GC)
C8H8ClNO2
MFCD00007165
2-methyl-3-nitrobenzyl chloride, 1-chloromethyl-2-methyl-3-nitrobenzene, alpha-chloro-2-methyl-3-nitrotoluene, 1-chloromethyl-2-methyl-3-nitro-benzene, 2-methyl-3-nitrobenzylchloride, benzene, 1-chloromethyl-2-methyl-3-nitro, acmc-1b2j5, ksc497o8t, 2-chloromethyl-6-nitrotoluene
CC1=C(CCl)C=CC=C1[N+]([O-])=O
185.61
185.61
97%
96.0
>109°C
Glass Bottle
CH3C6H3(NO2)CH2Cl
1g
XZNDXQGZPOZITR-UHFFFAOYSA-N
1-(chloromethyl)-2-methyl-3-nitrobenzene
108947
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 262-249-3

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only