missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-3-nitrophenylacetic acid, 96%, Thermo Scientific Chemicals

$44.67 - $183.03

Chemical Identifiers

CAS 23876-15-5
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD07782132
InChI Key LWIOFILTAJJDLA-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrophenylacetic acid, 2-methyl-3-nitrophenyl acetic acid, 2-2-methyl-3-nitrophenyl acetic acid, 2-methyl-3-nitro-benzeneacetic acid, 3-nitro-o-tolyl acetic acid, benzeneacetic acid, 2-methyl-3-nitro, 2-methyl-3-nitrophenylaceticacid, 2-methyl-3-nitrobenzeneacetic acid, 2-methyl-3-nitro phenyl acetic acid, pubchem18230
PubChem CID 13530897
SMILES CC1=C(C=CC=C1[N+](=O)[O-])CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6481006
View Documents
Thermo Scientific Chemicals
H6481006
5 g
Each for $44.67
 
AAH6481014
View Documents
Thermo Scientific Chemicals
H6481014
25 g
Each for $183.03
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23876-15-5
195.174
LWIOFILTAJJDLA-UHFFFAOYSA-N
13530897
C9H9NO4
MFCD07782132
2-methyl-3-nitrophenylacetic acid, 2-methyl-3-nitrophenyl acetic acid, 2-2-methyl-3-nitrophenyl acetic acid, 2-methyl-3-nitro-benzeneacetic acid, 3-nitro-o-tolyl acetic acid, benzeneacetic acid, 2-methyl-3-nitro, 2-methyl-3-nitrophenylaceticacid, 2-methyl-3-nitrobenzeneacetic acid, 2-methyl-3-nitro phenyl acetic acid, pubchem18230
CC1=C(C=CC=C1[N+](=O)[O-])CC(=O)O

Specifications

23876-15-5
C9H9NO4
5 g
2-methyl-3-nitrophenylacetic acid, 2-methyl-3-nitrophenyl acetic acid, 2-2-methyl-3-nitrophenyl acetic acid, 2-methyl-3-nitro-benzeneacetic acid, 3-nitro-o-tolyl acetic acid, benzeneacetic acid, 2-methyl-3-nitro, 2-methyl-3-nitrophenylaceticacid, 2-methyl-3-nitrobenzeneacetic acid, 2-methyl-3-nitro phenyl acetic acid, pubchem18230
CC1=C(C=CC=C1[N+](=O)[O-])CC(=O)O
13530897
96%
2-Methyl-3-nitrophenylacetic acid
132°C to 136°C
MFCD07782132
2581973
LWIOFILTAJJDLA-UHFFFAOYSA-N
195.174
195.17
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only