missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-4-nitroaniline 98.0+%, TCI America™
$44.73 - $493.97
Chemical Identifiers
| CAS | 99-52-5 |
|---|---|
| Molecular Formula | C7H8N2O2 |
| Molecular Weight (g/mol) | 152.15 |
| MDL Number | MFCD00007734 |
| InChI Key | XTTIQGSLJBWVIV-UHFFFAOYSA-N |
| Synonym | 2-amino-5-nitrotoluene, 4-nitro-o-toluidine, 5-nitro-2-aminotoluene, p-nitro-o-toluidine, benzenamine, 2-methyl-4-nitro, fast red rl base, 1-amino-2-methyl-4-nitrobenzene, diabase red rl, ansibases red rl, devol red rl |
| PubChem CID | 7441 |
| IUPAC Name | 2-methyl-4-nitroaniline |
| SMILES | CC1=CC(=CC=C1N)[N+]([O-])=O |
Chemical Identifiers
| 99-52-5 | |
| 152.15 | |
| XTTIQGSLJBWVIV-UHFFFAOYSA-N | |
| 7441 | |
| CC1=CC(=CC=C1N)[N+]([O-])=O |
| C7H8N2O2 | |
| MFCD00007734 | |
| 2-amino-5-nitrotoluene, 4-nitro-o-toluidine, 5-nitro-2-aminotoluene, p-nitro-o-toluidine, benzenamine, 2-methyl-4-nitro, fast red rl base, 1-amino-2-methyl-4-nitrobenzene, diabase red rl, ansibases red rl, devol red rl | |
| 2-methyl-4-nitroaniline |
Specifications
| 99-52-5 | |
| Yellow | |
| MFCD00007734 | |
| 2660 | |
| XTTIQGSLJBWVIV-UHFFFAOYSA-N | |
| 2-methyl-4-nitroaniline | |
| 7441 | |
| ≥98.0% (GC) | |
| 2-Methyl-4-nitroaniline |
| 131°C | |
| C7H8N2O2 | |
| 25 g | |
| 2-amino-5-nitrotoluene, 4-nitro-o-toluidine, 5-nitro-2-aminotoluene, p-nitro-o-toluidine, benzenamine, 2-methyl-4-nitro, fast red rl base, 1-amino-2-methyl-4-nitrobenzene, diabase red rl, ansibases red rl, devol red rl | |
| CC1=CC(=CC=C1N)[N+]([O-])=O | |
| 152.15 | |
| 152.15 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0401
RTECSNumber : XU8210000
TSCA : Yes