missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-4-phenyl-3-butyn-2-ol 98.0+%, TCI America™

$118.56 - $118.56

Chemical Identifiers

CAS 1719-19-3
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.22
MDL Number MFCD00041572
InChI Key FUPXYICBZMASCM-UHFFFAOYSA-N
Synonym 2-Hydroxy-2-methyl-4-phenyl-3-butyne
PubChem CID 288327
IUPAC Name 2-methyl-4-phenylbut-3-yn-2-ol
SMILES CC(C)(O)C#CC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M14151G
View Documents
TCI America
M14151G
1 g
Each for $118.56
 
Description

Description

Specifications

Chemical Identifiers

1719-19-3
160.22
FUPXYICBZMASCM-UHFFFAOYSA-N
288327
CC(C)(O)C#CC1=CC=CC=C1
C11H12O
MFCD00041572
2-Hydroxy-2-methyl-4-phenyl-3-butyne
2-methyl-4-phenylbut-3-yn-2-ol

Specifications

1719-19-3
White-Yellow
MFCD00041572
2-Hydroxy-2-methyl-4-phenyl-3-butyne
CC(C)(O)C#CC1=CC=CC=C1
160.22
160.22
Crystalline Powder
54°C
C11H12O
1 g
FUPXYICBZMASCM-UHFFFAOYSA-N
2-methyl-4-phenylbut-3-yn-2-ol
288327
≥98.0% (GC)
2-Methyl-4-phenyl-3-butyn-2-ol
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'