missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-4-trimethylsilyl-1-buten-3-yne, 97%, Thermo Scientific Chemicals
$81.63 - $865.60
Chemical Identifiers
CAS | 18387-60-5 |
---|---|
Molecular Formula | C8H14Si |
Molecular Weight (g/mol) | 138.285 |
MDL Number | MFCD00190206 |
InChI Key | HRGBALJHGYAWBL-UHFFFAOYSA-N |
Synonym | 2-methyl-4-trimethylsilyl-1-buten-3-yne, 2-methyl-4-trimethylsilylbut-1-en-3-yne, acmc-1cdn8, trimethylsilylisopropenylacetylene, trimethyl 3-methylbut-3-en-1-ynyl silane, 3-methyl-3-butene-1-ynyl trimethylsilane, trimethyl 3-methyl-3-buten-1-ynyl silane #, trimethyl 3-methylbut-3-en-1-yn-1-yl silane, silane,trimethyl 3-methyl-3-buten-1-yn-1-yl, 2-methyl-4-trimethylsilyl-1-buten-3-yne, |
PubChem CID | 579646 |
IUPAC Name | trimethyl(3-methylbut-3-en-1-ynyl)silane |
SMILES | CC(=C)C#C[Si](C)(C)C |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18387-60-5 | |
138.285 | |
HRGBALJHGYAWBL-UHFFFAOYSA-N | |
579646 | |
CC(=C)C#C[Si](C)(C)C |
C8H14Si | |
MFCD00190206 | |
2-methyl-4-trimethylsilyl-1-buten-3-yne, 2-methyl-4-trimethylsilylbut-1-en-3-yne, acmc-1cdn8, trimethylsilylisopropenylacetylene, trimethyl 3-methylbut-3-en-1-ynyl silane, 3-methyl-3-butene-1-ynyl trimethylsilane, trimethyl 3-methyl-3-buten-1-ynyl silane #, trimethyl 3-methylbut-3-en-1-yn-1-yl silane, silane,trimethyl 3-methyl-3-buten-1-yn-1-yl, 2-methyl-4-trimethylsilyl-1-buten-3-yne, | |
trimethyl(3-methylbut-3-en-1-ynyl)silane |
Specifications
18387-60-5 | |
32°C (15 mmHg) | |
MFCD00190206 | |
UN1993 | |
HRGBALJHGYAWBL-UHFFFAOYSA-N | |
trimethyl(3-methylbut-3-en-1-ynyl)silane | |
579646 | |
97% | |
2-Methyl-4-trimethylsilyl-1-buten-3-yne |
0.778 g/mL | |
C8H14Si | |
2 g | |
2-methyl-4-trimethylsilyl-1-buten-3-yne, 2-methyl-4-trimethylsilylbut-1-en-3-yne, acmc-1cdn8, trimethylsilylisopropenylacetylene, trimethyl 3-methylbut-3-en-1-ynyl silane, 3-methyl-3-butene-1-ynyl trimethylsilane, trimethyl 3-methyl-3-buten-1-ynyl silane #, trimethyl 3-methylbut-3-en-1-yn-1-yl silane, silane,trimethyl 3-methyl-3-buten-1-yn-1-yl, 2-methyl-4-trimethylsilyl-1-buten-3-yne, | |
CC(=C)C#C[Si](C)(C)C | |
138.285 | |
138.28 | |
Liquid |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only