Learn More
2-Methyl-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals
$120.23 - $463.66
Chemical Identifiers
CAS | 89976-12-5 |
---|---|
Molecular Formula | C8H6F3NO2 |
Molecular Weight (g/mol) | 205.136 |
MDL Number | MFCD01631684 |
InChI Key | SVQCVQCIZWSPPX-UHFFFAOYSA-N |
Synonym | 2-methyl-5-nitrobenzotrifluoride, 1-methyl-4-nitro-2-trifluoromethyl benzene, 2-trifluoromethyl-4-nitrotoluene, 4-nitro-2-trifluoromethyl toluene, 1-methyl-4-nitro-2-trifluoromethyl-benzene, benzene, 1-methyl-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-methylnitrobenzene, 2-trifluoromethyl-1-methyl-4-nitrobenzene, pubchem4444 |
PubChem CID | 2775447 |
IUPAC Name | 1-methyl-4-nitro-2-(trifluoromethyl)benzene |
SMILES | CC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6492406
|
Thermo Scientific Chemicals
H6492406 |
5 g |
Each for $120.23
|
|
|||||
AAH6492414
|
Thermo Scientific Chemicals
H6492414 |
25 g |
Each for $463.66
|
|
|||||
Description
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
Chemical Identifiers
89976-12-5 | |
205.136 | |
SVQCVQCIZWSPPX-UHFFFAOYSA-N | |
2775447 | |
CC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F |
C8H6F3NO2 | |
MFCD01631684 | |
2-methyl-5-nitrobenzotrifluoride, 1-methyl-4-nitro-2-trifluoromethyl benzene, 2-trifluoromethyl-4-nitrotoluene, 4-nitro-2-trifluoromethyl toluene, 1-methyl-4-nitro-2-trifluoromethyl-benzene, benzene, 1-methyl-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-methylnitrobenzene, 2-trifluoromethyl-1-methyl-4-nitrobenzene, pubchem4444 | |
1-methyl-4-nitro-2-(trifluoromethyl)benzene |
Specifications
89976-12-5 | |
C8H6F3NO2 | |
5 g | |
2506013 | |
2-methyl-5-nitrobenzotrifluoride, 1-methyl-4-nitro-2-trifluoromethyl benzene, 2-trifluoromethyl-4-nitrotoluene, 4-nitro-2-trifluoromethyl toluene, 1-methyl-4-nitro-2-trifluoromethyl-benzene, benzene, 1-methyl-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-methylnitrobenzene, 2-trifluoromethyl-1-methyl-4-nitrobenzene, pubchem4444 | |
SVQCVQCIZWSPPX-UHFFFAOYSA-N | |
1-methyl-4-nitro-2-(trifluoromethyl)benzene | |
2775447 | |
98% | |
2-Methyl-5-nitrobenzotrifluoride |
White | |
MFCD01631684 | |
UN2811 | |
Moisture sensitive | |
Slightly soluble in water. | |
CC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F | |
205.136 | |
205.14 | |
Solid |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only