missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-(trimethylsilyl)phenyl Trifluoromethanesulfonate 96.0+%, TCI America™
$380.27 - $1453.97
Chemical Identifiers
| CAS | 556812-44-3 |
|---|---|
| Molecular Formula | C11H15F3O3SSi |
| Molecular Weight (g/mol) | 312.38 |
| MDL Number | MFCD10566915 |
| InChI Key | ZCWWZFXNICWDIB-UHFFFAOYSA-N |
| Synonym | 2-Methyl-6-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 2-Methyl-6-(trimethylsilyl)phenyl Ester, 6-(Trimethylsilyl)-o-tolyl Trifluoromethanesulfonate |
| PubChem CID | 12182646 |
| IUPAC Name | 2-methyl-6-(trimethylsilyl)phenyl trifluoromethanesulfonate |
| SMILES | CC1=C(OS(=O)(=O)C(F)(F)F)C(=CC=C1)[Si](C)(C)C |
Chemical Identifiers
| 556812-44-3 | |
| 312.38 | |
| ZCWWZFXNICWDIB-UHFFFAOYSA-N | |
| 12182646 | |
| CC1=C(OS(=O)(=O)C(F)(F)F)C(=CC=C1)[Si](C)(C)C |
| C11H15F3O3SSi | |
| MFCD10566915 | |
| 2-Methyl-6-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 2-Methyl-6-(trimethylsilyl)phenyl Ester, 6-(Trimethylsilyl)-o-tolyl Trifluoromethanesulfonate | |
| 2-methyl-6-(trimethylsilyl)phenyl trifluoromethanesulfonate |
Specifications
| 556812-44-3 | |
| C11H15F3O3SSi | |
| 1 g | |
| 2-Methyl-6-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 2-Methyl-6-(trimethylsilyl)phenyl Ester, 6-(Trimethylsilyl)-o-tolyl Trifluoromethanesulfonate | |
| CC1=C(OS(=O)(=O)C(F)(F)F)C(=CC=C1)[Si](C)(C)C | |
| 312.38 | |
| 312.38 | |
| Liquid |
| Yellow | |
| MFCD10566915 | |
| 3265 | |
| ZCWWZFXNICWDIB-UHFFFAOYSA-N | |
| 2-methyl-6-(trimethylsilyl)phenyl trifluoromethanesulfonate | |
| 12182646 | |
| ≥96.0% (GC) | |
| 2-Methyl-6-(trimethylsilyl)phenyl Trifluoromethanesulfonate |
Safety and Handling
TSCA : No