missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-8-nitroquinoline 98.0+%, TCI America™
$169.63 - $466.02
Chemical Identifiers
| CAS | 881-07-2 |
|---|---|
| Molecular Formula | C10H8N2O2 |
| Molecular Weight (g/mol) | 188.19 |
| MDL Number | MFCD00006764 |
| InChI Key | UHPGVDHXHDPYQP-UHFFFAOYSA-N |
| PubChem CID | 13433 |
| IUPAC Name | 2-methyl-8-nitroquinoline |
| SMILES | CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O |
Chemical Identifiers
| 881-07-2 | |
| 188.19 | |
| UHPGVDHXHDPYQP-UHFFFAOYSA-N | |
| 2-methyl-8-nitroquinoline |
| C10H8N2O2 | |
| MFCD00006764 | |
| 13433 | |
| CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O |
Specifications
| 881-07-2 | |
| Yellow | |
| MFCD00006764 | |
| 8-nitroquinaldine, quinaldine, 8-nitro, quinoline, 2-methyl-8-nitro, unii-xrv949auqz, xrv949auqz, acmc-209qr6, 2-methyl-8-nitro-quinoline, 5-20-07-00387 beilstein handbook reference, quinoline,2-methyl-8-nitro, uhpgvdhxhdpyqp-uhfffaoysa | |
| CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O | |
| 188.19 | |
| 188.19 | |
| Crystalline Powder |
| 138°C | |
| C10H8N2O2 | |
| 5 g | |
| UHPGVDHXHDPYQP-UHFFFAOYSA-N | |
| 2-methyl-8-nitroquinoline | |
| 13433 | |
| ≥98.0% (GC,T) | |
| 2-Methyl-8-nitroquinoline |
Safety and Handling
RTECSNumber : UZ9830000
TSCA : No