missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H57776MD

 View more versions of this product

Catalog No. AAH57776MD


Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (1.00E+06 mg/L).

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
Specifications

Specifications

2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide
C7H13NO2S
250 mg
Soluble in water. (1.00E+06mg/L).
CC(C)(C)S(=O)N=C1COC1
175.246
175.25
1158098-73-7
MFCD14702522
2-methyl-n-oxetan-3-ylidene propane-2-sulfinamide, 2-methyl-n-3-oxetanylidene-2-propanesulfinamide, 2-methyl-n-3-oxetanylidene propane-2-sulfinamide, 3-tert-butylsulfinylmethylene oxetane, n-tert-butylsulfinyl oxetane-3-imine, 2-methyl-n-oxetan-3-ylidene-propane-2-sulfinamide, 2-methyl-n-oxetan-3-ylidene ;propane-2-sulfinamide, 2-methyl-propane-2-sulfinic acid oxetan-3-ylideneamide, 2-methylpropane-2-sulfinic acid oxetan-3-ylideneamide
VKUZMNXQGKBLHN-UHFFFAOYSA-N
2-methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide
46839995
95%

Chemical Identifiers

1158098-73-7
175.246
VKUZMNXQGKBLHN-UHFFFAOYSA-N
46839995
CC(C)(C)S(=O)N=C1COC1
C7H13NO2S
MFCD14702522
2-methyl-n-oxetan-3-ylidene propane-2-sulfinamide, 2-methyl-n-3-oxetanylidene-2-propanesulfinamide, 2-methyl-n-3-oxetanylidene propane-2-sulfinamide, 3-tert-butylsulfinylmethylene oxetane, n-tert-butylsulfinyl oxetane-3-imine, 2-methyl-n-oxetan-3-ylidene-propane-2-sulfinamide, 2-methyl-n-oxetan-3-ylidene ;propane-2-sulfinamide, 2-methyl-propane-2-sulfinic acid oxetan-3-ylideneamide, 2-methylpropane-2-sulfinic acid oxetan-3-ylideneamide
2-methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only