Learn More
2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H57776ME
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water. (1.00E+06 mg/L).
Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
Specifications
2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide | |
C7H13NO2S | |
500 mg | |
Soluble in water. (1.00E+06mg/L). | |
CC(C)(C)S(=O)N=C1COC1 | |
175.246 | |
175.25 |
1158098-73-7 | |
MFCD14702522 | |
2-methyl-n-oxetan-3-ylidene propane-2-sulfinamide, 2-methyl-n-3-oxetanylidene-2-propanesulfinamide, 2-methyl-n-3-oxetanylidene propane-2-sulfinamide, 3-tert-butylsulfinylmethylene oxetane, n-tert-butylsulfinyl oxetane-3-imine, 2-methyl-n-oxetan-3-ylidene-propane-2-sulfinamide, 2-methyl-n-oxetan-3-ylidene ;propane-2-sulfinamide, 2-methyl-propane-2-sulfinic acid oxetan-3-ylideneamide, 2-methylpropane-2-sulfinic acid oxetan-3-ylideneamide | |
VKUZMNXQGKBLHN-UHFFFAOYSA-N | |
2-methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide | |
46839995 | |
95% |
Chemical Identifiers
1158098-73-7 | |
175.246 | |
VKUZMNXQGKBLHN-UHFFFAOYSA-N | |
46839995 | |
CC(C)(C)S(=O)N=C1COC1 |
C7H13NO2S | |
MFCD14702522 | |
2-methyl-n-oxetan-3-ylidene propane-2-sulfinamide, 2-methyl-n-3-oxetanylidene-2-propanesulfinamide, 2-methyl-n-3-oxetanylidene propane-2-sulfinamide, 3-tert-butylsulfinylmethylene oxetane, n-tert-butylsulfinyl oxetane-3-imine, 2-methyl-n-oxetan-3-ylidene-propane-2-sulfinamide, 2-methyl-n-oxetan-3-ylidene ;propane-2-sulfinamide, 2-methyl-propane-2-sulfinic acid oxetan-3-ylideneamide, 2-methylpropane-2-sulfinic acid oxetan-3-ylideneamide | |
2-methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only