missing translation for 'onlineSavingsMsg'
Learn More

2-Methylanthraquinone, 97%

CAS: 84-54-8 | C15H10O2 | 222.243 g/mol

$60.19 - $559.60

Chemical Identifiers

CAS 84-54-8
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.243
MDL Number MFCD00001235
InChI Key NJWGQARXZDRHCD-UHFFFAOYSA-N
Synonym 2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
PubChem CID 6773
ChEBI CHEBI:9427
IUPAC Name 2-methylanthracene-9,10-dione
SMILES CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1450218
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
A1450218
50 g
Each for $60.19
Only null left
 
AAA1450230
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
A1450230
250 g
Each for $174.60
Only null left
 
AAA145020B
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
A145020B
1000 g
Each for $559.60
Only null left
 
Description

Description

2-Methylanthraquinone is used as a pharmaceutical intermediate. It is used in smog dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylanthraquinone is used as a pharmaceutical intermediate. It is used in smog dyes.

Solubility
Soluble in water 1.234 mg/L @ 25°C.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

84-54-8
222.243
NJWGQARXZDRHCD-UHFFFAOYSA-N
6773
2-methylanthracene-9,10-dione
C15H10O2
MFCD00001235
2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
CHEBI:9427
CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O

Specifications

84-54-8
209°C (408°F)
MFCD00001235
2050523
2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
NJWGQARXZDRHCD-UHFFFAOYSA-N
2-methylanthracene-9,10-dione
6773
222.24
2-Methylanthraquinone
170°C to 174°C
C15H10O2
50 g
14,6021
Soluble in water 1.234mg/L @ 25°C.
CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
222.243
CHEBI:9427
97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-539-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only