missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methylbenzoic Anhydride 98.0+%, TCI America™
$209.35 - $626.30
Chemical Identifiers
CAS | 607-86-3 |
---|---|
Molecular Formula | C16H14O3 |
Molecular Weight (g/mol) | 254.29 |
MDL Number | MFCD00048077 |
InChI Key | YLBSXJWDERHYFY-UHFFFAOYSA-N |
PubChem CID | 136405 |
IUPAC Name | 2-methylbenzoyl 2-methylbenzoate |
SMILES | CC1=CC=CC=C1C(=O)OC(=O)C1=CC=CC=C1C |
Chemical Identifiers
607-86-3 | |
254.29 | |
YLBSXJWDERHYFY-UHFFFAOYSA-N | |
2-methylbenzoyl 2-methylbenzoate |
C16H14O3 | |
MFCD00048077 | |
136405 | |
CC1=CC=CC=C1C(=O)OC(=O)C1=CC=CC=C1C |
Specifications
607-86-3 | |
White | |
C16H14O3 | |
1 g | |
CC1=CC=CC=C1C(=O)OC(=O)C1=CC=CC=C1C | |
254.29 | |
254.29 | |
Crystalline Powder |
39°C | |
221°C | |
MFCD00048077 | |
YLBSXJWDERHYFY-UHFFFAOYSA-N | |
2-methylbenzoyl 2-methylbenzoate | |
136405 | |
≥98.0% (GC,T) | |
2-Methylbenzoic Anhydride |
Safety and Handling
TSCA : No