missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-,Methylbutane, MilliporeSigma™
Supplier: Technidata MX07601
Specifications
2-, Methylbutane | |
-16°C | |
28°C (1013 hPa) | |
−57°C | |
Metal Can | |
(CH3)2CHCH2CH3 | |
4 L | |
QWTDNUCVQCZILF-UHFFFAOYSA-N | |
2-methylbutane | |
6556 | |
72.15 |
78-78-4 | |
0.62g/cm3 (20°C) | |
1.3 to 7.6%(V) | |
420°C | |
C5H12 | |
MFCD00009338 | |
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
CCC(C)C | |
72.15 | |
CHEBI:30362 | |
800 hPa (20°C) |
Safety and Handling
Recommended Storage : Store below +15°C