missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutyraldehyde, 95%

CAS: 96-17-3 | C5H10O | 86.13 g/mol

$220.66 - $220.66

Chemical Identifiers

CAS 96-17-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00006984
InChI Key BYGQBDHUGHBGMD-UHFFFAOYNA-N
Synonym 2-methylbutyraldehyde, butanal, 2-methyl, 2-formylbutane, 2-methylbutyric aldehyde, methylethylacetaldehyde, 2-ethylpropanal, 2-methyl-1-butanal, butyraldehyde, 2-methyl, acetaldehyde, ethylmethyl, alpha-methylbutanal
PubChem CID 7284
ChEBI CHEBI:16182
IUPAC Name 2-methylbutanal
SMILES CCC(C)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1965022
View Documents
Thermo Scientific Chemicals
A1965022
100 g
Each for $220.66
Only null left
 
Description

Description

2-Methylbutyraldehyde is used for lift in woody and cocoa topnote complexes. And also used in cocoa, coffee, wine and nut flavors for lift.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbutyraldehyde is used for lift in woody and cocoa topnote complexes. And also used in cocoa, coffee, wine and nut flavors for lift.

Solubility
Soluble in water, ether, and alcohol.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

96-17-3
86.13
BYGQBDHUGHBGMD-UHFFFAOYNA-N
7284
2-methylbutanal
C5H10O
MFCD00006984
2-methylbutyraldehyde, butanal, 2-methyl, 2-formylbutane, 2-methylbutyric aldehyde, methylethylacetaldehyde, 2-ethylpropanal, 2-methyl-1-butanal, butyraldehyde, 2-methyl, acetaldehyde, ethylmethyl, alpha-methylbutanal
CHEBI:16182
CCC(C)C=O

Specifications

96-17-3
94°C to 96°C
C5H10O
MFCD00006984
UN1989
Air Sensitive
Soluble in water,ether,and alcohol.
CCC(C)C=O
86.13
CHEBI:16182
95%
0.806
4°C (39°F)
1.3919
100 g
1633540
2-methylbutyraldehyde, butanal, 2-methyl, 2-formylbutane, 2-methylbutyric aldehyde, methylethylacetaldehyde, 2-ethylpropanal, 2-methyl-1-butanal, butyraldehyde, 2-methyl, acetaldehyde, ethylmethyl, alpha-methylbutanal
BYGQBDHUGHBGMD-UHFFFAOYNA-N
2-methylbutanal
7284
86.13
2-Methylbutyraldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c

H225-H317-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.

EINECSNumber : 202-485-6

RTECSNumber : ES3400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only